Learn More
Cdc37L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531292
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (95%), Rhesus Monkey (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008]
Spezifikation
Cdc37L1 | |
Polyclonal | |
Unconjugated | |
CDC37L1 | |
2700033A15Rik; BB159850; CDC37 cell division cycle 37 homolog-like 1; CDC37B; Cdc37l; Cdc37l1; cell division cycle 37 homolog-like 1; cell division cycle 37 like 1; cell division cycle 37-like 1; HARC; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55664 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q7L3B6 | |
CDC37L1 | |
Recombinant fragment corresponding to a region within amino acids 18 and 249 of Human CDC37L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.