Learn More
NFE2L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534837
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (96%), Chicken (87%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for 'nuclear respiratory factor 1' which has an official symbol of NRF1.
Spezifikation
NFE2L1 | |
Polyclonal | |
Unconjugated | |
NFE2L1 | |
AA408798; AW212678; bZIP protein; endoplasmic reticulum membrane sensor NFE2L1; HBZ17; Lcrf1; LCR-F1; locus control region-factor 1; Nfe2l1; NF-E2-related factor 1; NFE2-related factor 1; Nrf1; nuclear factor (erythroid-derived 2)-like 1; nuclear factor erythroid 2-like 1; nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2 like 1; nuclear factor, erythroid 2-like 1; nuclear factor, erythroid derived 2, like 1; nuclear factor, erythroid derived 2,-like 1; Protein NRF1, p110 form; Protein NRF1, p120 form; TCF11; TCF-11; transc; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1; Transcription factor NRF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4779 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.51 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q14494 | |
NFE2L1 | |
Recombinant fragment corresponding to a region within amino acids 495 and 745 of Nrf1 (Uniprot ID#Q14494) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.