Learn More
Hamilton™ 5 mL 1000 Series Gas Tight Syringe
The 1000 Series syringes offer a mid-volume Gas Tight solution.
Marke: Hamilton™ CAL81601
Beschreibung
- This syringe series boasts a variety of termination and needle options, providing exceptional versatility and adaptability.
- Dispenses volumes up to 5 mL.
Spezifikation
Nicht autoklavierbar | |
22 G | |
0,0161 in. | |
0,0283 in. | |
10.3 mm | |
0.72 mm | |
239 °F | |
50 °F | |
10 °C | |
5 mL | |
Spitz, schräg, gebogen, nicht stanzend |
PTFE | |
0.406 Zoll | |
0.41 mm | |
0.530 in. | |
13.5 mm | |
2 in. | |
115 °C | |
1005 SL | |
1 | |
51 mm | |
Probensperre |