missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6 Antibody, R&D Systems™
Recombinant Monoclonal Antibody
Marke: R&D Systems MAB2061R-01M
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.
Spezifikation
IL-6 | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3569 | |
Human | |
Purified |
Flow Cytometry, Neutralization | |
1936R | |
Neutralization 8.00-80.0 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
P05231 | |
IL6 | |
E. coli-derived recombinant human IL-6 Val30-Met212 Accession No. P05231 | |
1 mg | |
Primary | |
Detects human IL-6 in direct ELISAs. Does not cross-react with recombinant IL-6 from mouse, rat, or pig. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Produktvorschläge
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Human IL-6 Antibody, R&D Systems™ > 1 mg; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur