missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD31/PECAM-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 27 publications
Marke: R&D Systems BBA7
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry, Knockout Validated.
Spezifikation
CD31/PECAM-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 μg/mL, Immunohistochemistry 5 to 25 μg/mL, Immunoprecipitation, Immunocytochemistry 8 to 25 μg/mL, Knockout Validated 8 to 25 μg/mL | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5175 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunoprecipitation, Immunocytochemistry, Immunoassay | |
9G11 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
PECAM1 | |
Activated HUVEC human umbilical vein endothelial cells | |
100 μg | |
Primary | |
Detects human CD31/PEACAM-1. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 was observed. | |
Human | |
Purified |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-4 of
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Human CD31/PECAM-1 Antibody, R&D Systems™ > 100ug; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur