missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse S100A6 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Marke: R&D Systems AF4584
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
S100A6 Polyclonal specifically detects S100A6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
Spezifikation
S100A6 | |
Polyclonal | |
Unconjugated | |
P14069 | |
S100A6 | |
E. coli-derived recombinant mouse S100A6 Met1-Lys89 Accession # P14069 | |
100 μg | |
Primary | |
Detects human and mouse S100A6 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse S100A1 and recombinant mouse S100A4 is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
2A9, CABP, CACY, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRA, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin) | |
Sheep | |
Affinity Purified | |
RUO | |
6277 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-4 of
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Human/Mouse S100A6 Antibody, R&D Systems™ > 100μg; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur