missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™ Product Code.: 10275244

2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™

Product Code. 10275244
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10275244

Brand: Thermo Scientific Maybridge CC17405DE

Request Bulk or Custom Format

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Chemical Identifiers

CAS 210832-85-2
Molecular Formula C12H14BrNO2
Molecular Weight (g/mol) 284.15
MDL Number MFCD03783555
InChI Key OUGMZFJPRSTGMJ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenoneShow MoreShow Less
PubChem CID 2795357
SMILES BrCC(=O)C1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 2-bromo-1-(4-morpholin-4-ylphenyl)ethanone
Formula Weight 284.15
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-morpholinophenyl)-1-ethanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.