Learn More
hnRNP UL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA561207
Benachrichtigungen:
Um den Rabatt von 33.33 % zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
Immunogen sequence: NESGYERRPL EMEQQQAYRP EMKTEMKQGA PTSFLPPEAS QLKPDRQQFQ SRKRPYEENR GRGYFEHRED RRGRSPQPPA EEDE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
Spezifikation
hnRNP UL1 | |
Polyclonal | |
Unconjugated | |
Hnrnpul1 | |
Adenovirus early region 1B-associated protein 5; E130317O14Rik; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1BAP5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U like 1; heterogeneous nuclear ribonucleoprotein U-like 1; heterogeneous nuclear ribonucleoprotein U-like protein 1; Hnrnpul; Hnrnpul1; HNRPUL1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein U-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUJ2 | |
Hnrnpul1 | |
Recombinant protein corresponding to Human hnRNP UL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Research Use Only