Learn More
Fresolimumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Marke: Invitrogen MA541794
Beschreibung
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Fresolimumab has been used in trials studying the treatment of Primary Brain Tumors, Metastatic Breast Cancer, Diffuse Systemic Sclerosis, Pleural Malignant Mesothelioma, and Primary Focal Segmental Glomerulosclerosis.
Spezifikation
Fresolimumab | |
Recombinant Monoclonal | |
Unconjugated | |
GC-1008 | |
Human TGFB/TGF beta | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1.89 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG4 κ |