missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-heptynoate, 98+%, Thermo Scientific™

Product Code. 11357825
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16930-95-3
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00015275
InChI Key GDCQMGYMFGPMCX-UHFFFAOYSA-N
Synonym ethyl 2-heptynoate, 2-heptynoic acid, ethyl ester, 2-heptynoic acid ethyl ester, acmc-1ca7r, ethyl-2-heptynoate, 2-heptynoic acid, ethylester, hept-2-ynoic acid ethyl ester
PubChem CID 85647
IUPAC Name ethyl hept-2-ynoate
SMILES CCCCC#CC(=O)OCC

Specifications

Density 0.916
Boiling Point 193°C to 194°C
Odor Recognizable
Linear Formula CH3(CH2)3C≡CCOOCH2CH3
Refractive Index 1.444
Quantity 1 g
Beilstein 1756846
Formula Weight 154.21
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-heptynoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-heptynoate, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.