missing translation for 'onlineSavingsMsg'
Learn More
3-(Pentafluorothio)phenol, 97%, Thermo Scientific™ Product Code.: 15466647

3-(Pentafluorothio)phenol, 97%, Thermo Scientific™

Product Code. 15466647
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15466647

Brand: Thermo Scientific Alfa Aesar H33787.06

Request Bulk or Custom Format

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-31-1
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788515
InChI Key KVOACEHJTNSNBD-UHFFFAOYSA-N
Synonym 3-pentafluorothio phenol, 3-pentafluorosulfanyl phenol, 3-pentafluoro-$l^ 6-sulfanyl phenol, 3-pentafluoro-??-sulfanyl phenol, 3-pentafluorosulfanylphenol, 3-pentafluorosulfur phenol, 3-hydroxyphenylpentafluorosulfur vi, 3-pentafluoro-lambda-sulfanyl phenol, 3-pentafluoro-lambda∼6∼-sulfanyl phenol, 3-pentakis fluoranyl-$l^ 6-sulfanyl phenolShow MoreShow Less
PubChem CID 2779201
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC(=CC=C1)S(F)(F)(F)(F)F

Specifications

Quantity 5 g
Formula Weight 220.16
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)phenol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10176704 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Pentafluorothio)phenol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.