Learn More
KW-2449, MedChemExpress
MedChemExpress KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.
Marke: Medchem Express HY-10339
Beschreibung
KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.Spezifikation
KW-2449 | |
1000669-72-6 | |
100 mg | |
C20H20N4O | |
DMSO : ≥ 50 mg/mL (150.42 mM) | |
332.4 | |
99.72% |
Powder | |
White | |
Research | |
Cancer-Kinase/protease | |
O=C(C1=CC=C(/C=C/C2=NNC3=C2C=CC=C3)C=C1)N4CCNCC4 | |
332.4 | |
Research |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für