Learn More
BUCHI FlashPure Chromatography Solid Loader
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
Marke:  BUCHI 148634332
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16340316
  • 136.00 € / 10 Stück
Verfügbar ab: 22-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Spezifikation
Spezifikation
Polypropylen
Fritten
10 bis 14 bar (145 bis 200 psi)
Empty Solid Loader
40 g
Name des Produkts
BUCHI FlashPure Chromatography Solid Loader > FlashPure Empty Solid Loader; Size: 40 g; Frits: Included; Quantity; 10 pcs