Learn More
CALCOCO2 Monoclonal Antibody (P1813), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA518293
Beschreibung
Recommended positive controls: Huh-7, Hep3B, HepG2, Jurkat, Raji, NCI-H929, Huh-7 (untreated), Huh-7 (3 μM Thapsigargin treatment for 12 hr), Huh-7 (3 μM Thapsigargin treatment for 24 hr), A.HepG2 (untreated), HepG2 (3 μM Thapsigargin treatment for 12 hr), HepG2 (3 μM Thapsigargin treatment for 24 hr). Predicted reactivity: Human (99%), Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CALCOCO2 Monoclonal antibody specifically detects CALCOCO2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation
CALCOCO2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
A2A6M5, Q13137 | |
CALCOCO2 | |
Recombinant fragment corresponding to a region within amino acids 174 and 429 of NDP52 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
P1813 | |
Unconjugated | |
CALCOCO2 | |
2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974 | |
Mouse | |
Protein G | |
RUO | |
10241, 76815 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Produktvorschläge
Customers who viewed this item also viewed.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.