Learn More
anti-MAPK14, MAX Protein Protein Interaction Antibody Pair, Abnova™
Marke: Abnova DI0533.1Set
Beinhaltet: MAPK14 rabbit purified polyclonal antibody (20μg); MAX mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Beschreibung
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK14 protein, and the other against the MAX protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spezifikation
MAPK14 | |
Monoclonal/Polyclonal | |
MAPK14 | |
MAPK14 | |
1 Set | |
Primary | |
Antibody pair set content: 1. MAPK14 rabbit purified polyclonal antibody (20 ug) 2. MAX mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA | |
Mouse, Rabbit | |
RUO | |
1432/4149 | |
Human | |
MAPK14 & MAX Protein Protein Interaction Antibody Pair |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.