missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Kinase 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552297
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: EKGQLVPLET VLDMLRDAMV AKVNTSKGFL IDGYPREVQQ GEEFERRIGQ PTLLLYVDAG PETMTQRLLK RGETSGRVDD NEETIKKRLE TYYKATEPVI AFYEKRGIVR KVNAEGS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 69%.
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.Spezifikation
Adenylate Kinase 1 | |
Polyclonal | |
Unconjugated | |
AK1 | |
adenylate kinase 1; adenylate kinase isoenzyme 1; adenylate kinase isoenzyme 1 {ECO:0000255; Adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255; AK 1; AK 1 {ECO:0000255; Ak1; Ak-1; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255; ATP-AMP transphosphorylase 1; ATP-AMP transphosphorylase 1 {ECO:0000255; B430205N08Rik; cytosolic adenylate kinase; HAMAP-Rule:MF_03171}; HTL-S-58j; I79_014777; Myokinase; myokinase {ECO:0000255; testis secretory sperm binding protein Li 58j | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11636, 203, 24183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00568, P39069, Q9R0Y5 | |
AK1 | |
Recombinant protein corresponding to Human Adenylate Kinase 1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-5 of 12
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Adenylate Kinase 1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur