CAS RN 10025-91-9
CAS RN 10025-91-9
Antimon(III)-chlorid, 99+ %, Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon-Trichlorid, 99.5 %, Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimontrichlorid, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, weiß, kristallin, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC-Name: Trichlorstiban SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, ACS, 99.0 % min., Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC-Name: Trichlorstiban SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, Puratronic™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, puriss p.a., ACS Reagenz, ≥99.0 % (RT), Honeywell™ Fluka™
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: Antimontrichlorid,Trichloro-Stibin,Antimon(V)-chlorid,Antimon(III)-chlorid,Stiban, Trichloro-,Antimonchlorid,Antimonbutter,Antimonlauge,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC-Name: Trichlorstiban SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimon(III)-chlorid, ≥99.0 %, ACS-Reagenz, Honeywell™ Fluka™
CAS: 10025-91-9 Summenformel: Cl3Sb Molekulargewicht (g/mol): 228.11 MDL-Nummer: MFCD00011212 InChI-Schlüssel: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: Antimontrichlorid,Trichloro-Stibin,Antimon(V)-chlorid,Antimon(III)-chlorid,Stiban, Trichloro-,Antimonchlorid,Antimonbutter,Antimonlauge,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC-Name: antimony(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]