Learn More
Thermo Scientific™ Capit-All Flex Automatischer Decapper
Der Thermo Scientific™ Capit-All™ Flex Automatische Decapper ist eine automatisierte Komplettlösung für das Verschließen und Öffnen von 48er- und 96er-Röhrchen-Racks.
Marke: Thermo Scientific™ 4120-FLEX
77135.00 EUR gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Beschreibung
Experience unparalleled efficiency with our advanced cryotube decapper system, designed to meet the needs of modern laboratories. Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats, this all-in-one instrument combines the functionality of four separate instruments, significantly reducing your laboratory's footprint and costs. Switch seamlessly between tube types without the hassle of adapters or part changes. The large 7 in. angled touchscreen, operable with or without gloves, ensures ease of use and visibility. A built-in drip tray acts as a barrier between caps and tubes to prevent cross-contamination and maintain the integrity of your samples. Our instrument includes tube verification to minimize errors and supports both full and partial racks, adapting to your specific needs. Whether used on the benchtop or an integrated system, it offers flexibility for your storage applications.
Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats Functionality of 4 instruments in one, reducing footprint and spend - Switch between tube types without the need of adapters or changing parts
- Large angled 7 in. touchscreen that can be used with or without gloves
- Built-in drip tray that acts as a barrier between caps and tubes to help prevent cross-contamination and maintain sample integrity
- Tube verification to prevent errors
- Works with full or partial racks of tubes
- Can be used for benchtop or integrated applications - fits inside a Biosafety Cabinet
Spezifikation
Matrix-Röhrchen | |
Automated Decapper | |
100/240 VAC |
96-Kanal | |
Touchscreen-Benutzeroberfläche | |
Ω |