Learn More
PCDHGA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552603
365.35 EUR gültig bis 2025-03-29
Benutzen Sie den Promotions-Code "24111" um Ihren Promotions-Preis zu erhalten.
Benachrichtigungen:
Um den Rabatt von 33.33 % zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
Immunogen sequence: PNTDWRFSQA QRPGTSGSQN GDDTGTWPNN QFDTEMLQAM ILASASEAAD GSSTLGGGAG TMGLSARYGP QFTLQHVPDY RQNVYIPGSN ATLTNAAGKR DGKAPAGGNG NKKKSGKKEK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 89%.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Spezifikation
PCDHGA1 | |
Polyclonal | |
Unconjugated | |
PCDHGA1 | |
PCDHGA1; PCDH-GAMMA-A1; protocadherin gamma a1; protocadherin gamma subfamily A, 1; protocadherin gamma-A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
553129, 56114, 93709 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5H4 | |
PCDHGA1 | |
Recombinant protein corresponding to Human PCDHGA1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Research Use Only