missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT6B-MORF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
193.30 € - 457.87 €
Spezifikation
Antigen | KAT6B-MORF |
---|---|
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Form | Purified |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18326236
|
Bio-Techne
NBP3-02990-20UL |
20 μL |
193.30 €
20 Mikroliter |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18361754
|
Bio-Techne
NBP3-02990-100UL |
100 μL |
457.87 €
100 Mikroliter |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
KAT6B-MORF Polyclonal specifically detects KAT6B-MORF in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
KAT6B-MORF | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
23522 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
DKFZp313G1618, FLJ90335, Histone acetyltransferase MORF, Histone acetyltransferase MOZ2, histone acetyltransferase MYST4, KAT6B, KIAA0383, Monocytic leukemia zinc finger protein-related factor, MORFMorf, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 4, MOZ2EC 2.3.1.48, MOZ-related factor, MYST histone acetyltransferase (monocytic leukemia) 4, MYST-4, qkf, querkopf | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human KAT6B-MORF (NP_001243398.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |