Learn More
EIF3A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26178-1-AP-150UL
Beschreibung
The German cDNA consortium.Spezifikation
EIF3A | |
Polyclonal | |
Unconjugated | |
EIF3A | |
eIF 3 theta, EIF3, eIF3 p167, eIF3 p170, eIF3 p180, eIF3 p185, eIF3 theta, EIF3A, EIF3S10, KIAA0139, P167, p180, p185, TIF32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8661 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14152 | |
EIF3A | |
EIF3A Fusion Protein Ag22515 | |
150 μL | |
Primary | |
Human | |
Antibody | |