Learn More
AGI-24512, MedChemExpress
MedChemExpress AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor, with an IC50 of 8 nM. AGI-24512 triggers DNA damage response. AGI-24512 can block proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer.
Marke: Medchem Express HY-112130
Beschreibung
AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor, with an IC50 of 8 nM. AGI-24512 triggers DNA damage response. AGI-24512 can block proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer.Spezifikation
AGI-24512 | |
Gray | |
C24H24N4O2 | |
DMSO : 125 mg/mL (312.13 mM; Need ultrasonic) | |
400.47 | |
99.48% | |
Solid |
2201066-53-5 | |
Research | |
50 mg | |
O=C1C(C2=CC=C(O)C=C2)=C(C)NC3=C(N4CCCCC4)C(C5=CC=CC=C5)=NN13 | |
400.47 | |
Research | |
Cancer-programmed cell death |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für