Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ MP-250D Series Digital Melting Point Apparatus
Accessories compatible with digital melting point apparatus.
Marke: Cole-Parmer™ 03010-69
Beschreibung
- 1 year warranty.
Spezifikation
Assay für PCB-Prozessoren | |
MP-250D Series Digital Melting Point Apparatus |