CAS RN 10540-29-1
CAS RN 10540-29-1
IUPAC Name:
(2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Synonyme:
tamoxifen tamoxifen preparation retaxim citofen tamofen tamax tamone terimon jenoxifen taxus
Molekulargewicht (g/mol):
371.52
Summenformel:
C26H29NO
InChi Key:
NKANXQFJJICGDU-QPLCGJKRSA-N
SMILES:
CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
1
–
6
von
6
Ergebnisse
1
Tamoxifen, 98 %, Thermo Scientific Chemicals
CAS: 10540-29-1 Summenformel: C26H29NO Molekulargewicht (g/mol): 371.52 MDL-Nummer: MFCD00010454 InChI-Schlüssel: NKANXQFJJICGDU-QPLCGJKRSA-N Synonym: tamoxifen,trans-tamoxifen,crisafeno,soltamox,tamoxifene,diemon,tamoxifeno,tamoxifenum,citofen,istubol PubChem CID: 2733526 ChEBI: CHEBI:41774 IUPAC-Name: (2-{4-[(1Z)-1,2-Diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamin SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
4
Tamoxifen, MP Biomedicals
CAS: 10540-29-1 Summenformel: C26H29NO Molekulargewicht (g/mol): 371.52 MDL-Nummer: MFCD00010454 InChI-Schlüssel: NKANXQFJJICGDU-QPLCGJKRSA-N SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
5
STEMCELL Technologies Tamoxifen
Estrogen signaling pathway modulator; Modulates selective estrogen receptor and estrogen-related receptor
6