CAS RN 10124-36-4
CAS RN 10124-36-4
Cadmiumsulfat, Thermo Scientific Chemicals
CAS: 10124-36-4 Summenformel: CdO4S Molekulargewicht (g/mol): 208.46 InChI-Schlüssel: QCUOBSQYDGUHHT-UHFFFAOYSA-L Synonym: cadmium sulfate,cadmium sulphate,cadmium monosulfate,sulfuric acid, cadmium salt 1:1,cadmium sulfuricum,cadmium sulfate 1:1,cadmium sulfate, tetrahydrate,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o PubChem CID: 24962 ChEBI: CHEBI:50292 IUPAC-Name: Cadmium(2+);Sulfat SMILES: [O-]S(=O)(=O)[O-].[Cd+2]
Cadmiumsulfat, ≥ 99 %, für Analysen, Thermo Scientific Chemicals
CAS: 10124-36-4 Summenformel: CdO4S Molekulargewicht (g/mol): 208.46 MDL-Nummer: MFCD00010923 InChI-Schlüssel: QCUOBSQYDGUHHT-UHFFFAOYSA-L Synonym: cadmium sulfate,cadmium sulphate,cadmium monosulfate,sulfuric acid, cadmium salt 1:1,cadmium sulfuricum,cadmium sulfate 1:1,cadmium sulfate, tetrahydrate,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o PubChem CID: 24962 ChEBI: CHEBI:50292 IUPAC-Name: Cadmium(2+);Sulfat SMILES: [O-]S(=O)(=O)[O-].[Cd+2]
Cadmiumsulfat, wasserfrei, ACS, > 99 %, Thermo Scientific Chemicals
CAS: 10124-36-4 Summenformel: CdO4S Molekulargewicht (g/mol): 208.47 MDL-Nummer: MFCD00010923 InChI-Schlüssel: QCUOBSQYDGUHHT-UHFFFAOYSA-L Synonym: cadmium sulfate,cadmium sulphate,cadmium monosulfate,sulfuric acid, cadmium salt 1:1,cadmium sulfuricum,cadmium sulfate 1:1,cadmium sulfate, tetrahydrate,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o PubChem CID: 24962 ChEBI: CHEBI:50292 IUPAC-Name: Cadmium(2+);Sulfat SMILES: [O-]S(=O)(=O)[O-].[Cd+2]
Cadmiumsulfat, ACS-Reagenz, ≥99 %, Honeywell™ Fluka™
CAS: 10124-36-4 Summenformel: CdO4S Molekulargewicht (g/mol): 208.47 MDL-Nummer: MFCD00010923 InChI-Schlüssel: QCUOBSQYDGUHHT-UHFFFAOYSA-L Synonym: Cadmiumsulfat,Cadmiummonosulfat,Schwefelsäure, Kadmiumsalz 1:1,Cadmiumsulfat 1:1,Cadmiumsulfat, Tetrahydrat,Cadmium2+-Sulfat,Caswell Nr. 136C,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o PubChem CID: 24962 ChEBI: CHEBI:50292 IUPAC-Name: Cadmium(2+);Sulfat SMILES: [O-]S(=O)(=O)[O-].[Cd+2]