missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Specific Gravity Hydrometer, Wide-Range
The Wide-Range Specific Gravity Hydrometer is ideal when you aren't sure of the approximate value of your liquid.
Marke: VEE GEE 6604-4
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Subdivisions: 0.005
- Approx. Length: 165 mm
- Calibrated for use at 15.5°C/60°F
Spezifikation
Specific Gravity Hydrometer | |
16.5 cm | |
15.6°C (60°F) |
0.005 | |
1.600 to 1.850 |