Learn More
Restek™ Rxi™-XLB GC Columns
General-purpose columns exhibiting extremely low bleed
Marke: Restek™ 43702
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
- Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
- Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Spezifikation
0.18 mm | |
Pestizidanalyse, PCB-Kongener-Analysen, PAHs Analysis | |
30°C to 340/360°C | |
20 m | |
Quarzglas |
Rxi™-XLB | |
0,18 μm | |
0.18 μm | |
Low polar |