Learn More
Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials.
Marke: Restek™ 24520
Weitere Details : Netto-Gewicht : 0.20000kg
Beschreibung
- Big mouth
- Pack of 100
Spezifikation
Polypropylen | |
Big Mouth without Spring | |
Fläschchen-Einsatz |
250 μl | |
100/Pk. |