Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
Fluted tip guides the capillary column into the jet
Marke: Restek™ 21620
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Threads specially coated for easy installation and removal
- Standard version undergoes special processing ensures the highest degree of cleanliness
- High-performance Siltek™ version is extremely inert for use with active compounds
Spezifikation
High-Performance Siltek Treated | |
JE | |
Agilent 6890/6850 |