missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Prolactin Antagonist Protein
A cDNA sequence encoding the Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP14010-EC-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Prolactin Antagonist Protein | |
Research Use Only | |
Ovine Prolactin Antagonist is devoid of agonistic activity and capable of inhibiting biological activity of Ovine Prolactin or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells. | |
Ovine | |
His | |
Greater than 99.0% as determined by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3. |