missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Streptokinase Protein
A cDNA sequence encoding the Streptokinase was constructed and used to recombinantly synthesize the protein.
339.00 € - 1110.00 €
Spezifikation
Name | Streptokinase Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The specific biological activity measured by the ability of fibrin lysis in agarose plate was found to be 80000U/mg. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16049441
|
enQuireBio™
QP10871-1,500,000IU |
1,500,000 IU |
1110.00 €
1 Stück |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915428
|
enQuireBio™
QP10871-250,000IU |
250,000 IU |
339.00 €
250000 Einheiten |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925428
|
enQuireBio™
QP10871-750,000IU |
750,000 IU |
600.00 €
750000 Einheiten |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
Streptokinase Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
IAGPEWLLDR PSVNNSQLVV SVAGTVEGTN QDISLKFFEI DLTSRPAHGG KTEQGLSPKS KLFATDSGAM PHKLEKADLL KAIQEQLIAN VHSNDDYFEV IDFASDATIT DRNGKVYFAD KDGSVTLPIQ PVQEFLLKGH VRVRPYKEKP VQNQAKSVDV EYTVQFTPLN PDDDFRPALK DTKLLKTLAI GDTITSQELL AQAQSILNKN HPGYTIYERD SSIVTHDNDI FRTILPMDQE FTYHVKNREQ AYRINKKSGL NEEINNTDLI SEKYYVLKKG EKPYDPFDRS HLKLFTIKYV DVNTNELLKS EQLLTASERN LDFRDLYDPR DKAKLLYNNL DAFGIMDYTL TGKVEDNHDD TNRIITVYMG KRPEGENASY HLAYDKDRYT EEEREVYSYL RYTGTPIPDN PNDK | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The specific biological activity measured by the ability of fibrin lysis in agarose plate was found to be 80000U/mg. | |
Other | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated (1 mg/ml) solution in PBS, pH 7.4. |