missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Streptokinase Protein

A cDNA sequence encoding the Streptokinase was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10871-250,000IU

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15915428

  • 339.00 € / 250000 Einheiten
Verfügbar ab: 22-10-2024
Sie sich ein, um den Lagerbestand zu sehen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

Streptokinase Protein
Research Use Only
The specific biological activity measured by the ability of fibrin lysis in agarose plate was found to be 80000U/mg.
Other
Untagged
Lyophilized from a 0.2microm filtered concentrated (1 mg/ml) solution in PBS, pH 7.4.
250,000 IU
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
IAGPEWLLDR PSVNNSQLVV SVAGTVEGTN QDISLKFFEI DLTSRPAHGG KTEQGLSPKS KLFATDSGAM PHKLEKADLL KAIQEQLIAN VHSNDDYFEV IDFASDATIT DRNGKVYFAD KDGSVTLPIQ PVQEFLLKGH VRVRPYKEKP VQNQAKSVDV EYTVQFTPLN PDDDFRPALK DTKLLKTLAI GDTITSQELL AQAQSILNKN HPGYTIYERD SSIVTHDNDI FRTILPMDQE FTYHVKNREQ AYRINKKSGL NEEINNTDLI SEKYYVLKKG EKPYDPFDRS HLKLFTIKYV DVNTNELLKS EQLLTASERN LDFRDLYDPR DKAKLLYNNL DAFGIMDYTL TGKVEDNHDD TNRIITVYMG KRPEGENASY HLAYDKDRYT EEEREVYSYL RYTGTPIPDN PNDK
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote