missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Importin subunit beta-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Importin subunit beta-1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6274-ec-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_032405.3 | |
Importin subunit beta-1 Protein | |
Research Use Only | |
Kpnb1 | |
Recombinant Protein | |
E. coli | |
MELITILEKT VSPDRLELEA AQKFLERAAV ENLPTFLVEL SRVLANPGNS QVARVAAGLQ IKNSLTSKDP DIKAQYQQRW LAIDANARRE VKNYVLQTLG TETYRPSSAS QCVAGIACAE IPVSQWPELI PQLVANVTNP NSTEHMKEST LEAIGYICQD IDPEQLQDKS NEILTAIIQG MRKEEPSNNV KLAATNALLN SLEFTKANFD KESERHFIMQ VVCEATQCPD TRVRVAALQN LVKIMSLYYQ YMETYMGPAL FAITIEAMKS DIDEVALQGI EFWSNVCDEE MDLAIEASEA AEQGRPPEHT SKFYAKGALQ YLVPILTQTL TKQDENDDDD DWNPCKAAGV CLMLLSTCCE DDIVPHVLPF IKEHIKNPDW RYRDAAVMAF GSILEGPEPN QLKPLVIQAM PTLIELMKDP SVVVRDTTAW TVGRICELLP EAAINDVYLA PLLQCLIEGL SAEPRVASNV CWAFSSLAEA AYEAADVADD QEEPATYCLS SSFELIVQKL LETTDRPDGH QNNLRSSAYE SLMEIVKNSA KDCYPAVQKT TLVIMERLQQ VLQMESHIQS TSDRIQFNDL QSLLCATLQN VLRKVQHQDA LQISDVVMAS LLRMFQSTAG SGGVQEDALM AVSTLVEVLG GEFLKYMEAF KPFLGIGLKN YAEYQVCLAA VGLVGDLCRA LQSNILPFCD EVMQLLLENL GNENVHRSVK PQILSVFGDI ALAIGGEFKK YLEVVLNTLQ QASQAQVDKS DFDMVDYLNE LRESCLEAYT GIVQGLKGDQ ENVHPDVMLV QPRVEFILSF IDHIAGDEDH TDGVVACAAG LIGDLCTAFG KDVLKLVEAR PMIHELLTEG RRSKTNKAKT LATWATKELR KLKNQA | |
Greater than 90% as determined by SDS-PAGE. |
16211 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |