missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cytochrome P450 2E1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 2E1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9094-ye-50ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_067257.1 | |
Cytochrome P450 2E1 Protein | |
Research Use Only | |
Cyp2e1 | |
Recombinant Protein | |
Yeast | |
AVLGITVALL VWIATLLLVS IWKQIYRSWN LPPGPFPIPF FGNIFQLDLK DIPKSLTKLA KRFGPVFTLH LGQRRIVVLH GYKAVKEVLL NHKNEFSGRG DIPVFQEYKN KGIIFNNGPT WKDVRRFSLS ILRDWGMGKQ GNEARIQREA HFLVEELKKT KGQPFDPTFL IGCAPCNVIA DILFNKRFDY DDKKCLELMS LFNENFYLLS TPWIQAYNYF SDYLQYLPGS HRKVMKNVSE IRQYTLGKAK EHLKSLDINC PRDVTDCLLI EMEKEKHSQE PMYTMENISV TLADLFFAGT ETTSTTLRYG LLILMKYPEI EEKLHEEIDR VIGPSRAPAV RDRMNMPYMD AVVHEIQRFI NLVPSNLPHE ATRDTVFRGY VIPKGTVVIP TLDSLLFDNY EFPDPETFKP EHFLNENGKF KYSDYFKAFS AGKRVCVGEG LARMELFLLL SAILQHFNLK SLVDPKDIDL SPVTIGFGSI PREFKLCVIP RS | |
Greater than 90% as determined by SDS-PAGE. |
13106 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |