missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF R alpha Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
362.00 €
Spezifikation
Gen-ID (Entrez) | 5156 |
---|---|
Molekulargewicht | 56 kDa |
Name | PDGFR alpha |
Menge | 50 μg |
Quelle | Mouse myeloma cell line,NS0-derived human PDGF R alpha protein Gln24-Glu524 |
Produktcode | Marke | Zusammensetzung | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Zusammensetzung | Preis | Menge & Verfügbarkeit | |||||
18384977
|
R&D Systems™
322-PR-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
362.00 €
50 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
The Recombinant Human PDGF R alpha Protein is derived from NS0. The Recombinant Human PDGF R alpha Protein has been validated for the following applications: Bioactivity.Spezifikation
5156 | |
PDGFR alpha | |
Mouse myeloma cell line,NS0-derived human PDGF R alpha protein Gln24-Glu524 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
56 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |