missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF R alpha Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 322-PR-050/CF
285.09 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human PDGF R alpha is immobilized at 2 μg/mL (100 μL/well), Recombinant Human PDGF-AB (Catalog # 222-AB ) binds with an ED50 of 2-12 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
56 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
5156 | |
PDGFR alpha | |
Mouse myeloma cell line,NS0-derived human PDGF R alpha protein Gln24-Glu524 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |