missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1E Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1E, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP8895-ec-100ug

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16077041

  • 554.00 € / 100 Mikrogramm

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Specifications

Specifications

NP_783316.2
Metallothionein-1E Protein
Research Use Only
MT1E
Recombinant Protein
E. coli
NCSCATGGSC TCAGSCKCKE CKCTSCKKSC CSCCPVGCAK CAQGCVCKGA SEKCSC
Greater than 90% as determined by SDS-PAGE.
4493
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Metallothionein-1E Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.