missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 nef Protein
A cDNA sequence encoding the HIV-1 nef was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP14022-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HIV-1 nef Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
kwskssv igwptvrerm rraepaadgv gaasqdlekh gaitssntaa tnadcawleaqeeeevgfpv tpqvplrpmt ykaavdlshf lkekgglegl ihsqrrqdil dlwiyhtqgyfpdwqnytpg pgirypltfg wcyklvpvep ekleeankge ntsllhpvsl hgmddperevlewrfdsrla | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
HIV-1 nef protein solution in 8M urea, 20mM Tris-HCl pH 8.0, 10mM b-mercaptoethanol. |