missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Glycolipid transfer Protein
A DNA sequence encoding the Bos taurus (Bovine) Glycolipid transfer protein, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6098-ec-10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_786993.1 | |
Glycolipid transfer Protein | |
Research Use Only | |
GLTP | |
Recombinant Protein | |
E. coli | |
ALLAEHLLRP LPADKQIETG PFLEAVSHLP PFFDCLGSPV FTPIKADISG NITKIKAVYD TNPTKFRTLQ NILEVEKEMY GAEWPKVGAT LALMWLKRGL RFIQVFLQSI CDGERDENHP NLIRVNATKA YEMALKKYHG WIVQKIFQAA LYAAPYKSDF LKALSKGQNV TEEECLEKVR LFLVNYTATI DVIYEMYTRM NAELNYKV | |
Greater than 90% as determined by SDS-PAGE. |
327678 | |
10 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |