missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein C inhibitor Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Spezifikation
Antigen | Protein C inhibitor |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16877203
|
Proteintech
10673-1-AP-20UL |
20 μL |
145.00 €
20 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
16867203
|
Proteintech
10673-1-AP-150UL |
150 μL |
410.00 €
150 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
Protein C inhibitor | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P05154 | |
5104 | |
Protein C inhibitor Fusion Protein Ag1078 | |
Primary | |
-20°C | |
SERPINA5 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PAI3, PCI, PLANH3, PROCI, Protein C inhibitor, SERPINA5 | |
SERPINA5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |