Learn More
Protein C inhibitor Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10673-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
Protein C inhibitor | |
Polyclonal | |
Unconjugated | |
SERPINA5 | |
PAI3, PCI, PLANH3, PROCI, Protein C inhibitor, SERPINA5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5104 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05154 | |
SERPINA5 | |
Protein C inhibitor Fusion Protein Ag1078 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |