missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ Staph Xtra Latex Kits
A rapid, sensitive method to distinguish Staphylococcus aureus and methicillin resistant Staphylococcus aureus (MRSA) from other species of Staphylococci
Marke: Pro-Lab Diagnostics™ PL.1080
Weitere Details : Netto-Gewicht : 0.20000kg
Beinhaltet:
- Staph Xtra Latex Reagent: 2 x 2.5mL (PL.1080), 2 x 7.5mL (PL.1081)
- Negative Control Latex Reagent: 5.0mL (PL.1080), 2 x 7.5mL (PL.1081)
- Agglutination Cards: 20 (PL.1080), 48 (PL.1081)
- Mixing Sticks: 200 (PL.1080), 300 (PL.1081)
Beschreibung
- Rapid clear results within 20 seconds
- Uses blue latex particles (CMMP) against a white background
Spezifikation
S. aureus | |
Staph Xtra Latex Kit | |
2 x 2.5mL Staph Xtra Latex reagent, 2.5mL negative control Latex reagent, 20 Agglutination cards, 200 mixing sticks | |
100 Tests |