missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fibrinogen from Bovine Plasma
Off-white shiny flaky powder
Marke: MP Biomedicals™ 0215112210
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Fibrinogen is blood protein that is involved in clotting and is converted to fibrin by thrombin. It consists of three non-identical pairs of disulfidebonded chains. The a-chain has an approximate molecular weight of 63.5 kDa, the b-chain 56 kDa, and the g-chain 47 kDa.
- Molecular weight: 340kDa
Cell Biology and Analysis, Molecular biology
Spezifikation
Cell Biology and Analysis, Molecular Biology | |
Fibrinogen from Bovine Plasma | |
Bovine Plasma | |
Powder |
340 | |
10g | |
0°C |