Learn More
Huber™ Minichiller
Click to view available options
Modell:
Mini-Chiller 300 W OLE
Minichiller 1000 OLE
Minichiller 300 OLE
Minichiller 300w-H OLE
Minichiller 600 OLE
Minichiller 600-H OLE
Kältemittel:
Kompakt
Natürliches Kältemittel
Packungsgröße:
1 Stück
Beschreibung
The compact Minichillers are an economic and environmentally friendly cooling solution for many laboratory applications. A calculation of water and waste water costs shows that a Minichiller can save up to 48,000 L of water in a working week and due to its low procurement price, return on investment is achieved within a very short time.
- The units have small footprints, only requiring 225 x 360 mm on the laboratory bench and are therefore suitable for cooling reactor blocks, vacuum pumps, rotary evaporators or heat exchangers.
- Minichillers allow stable application conditions due to constant values for temperature, pressure and throughflow quantities.
- They are also perfect for continuous operation at ambient temperatures up to + 40°C.
- Despite the economic purchase price the coolers are well equipped and have a large temperature indicator, fill level indicator, as well as status LEDs for the pump, cooling and heating.
- A 1 kW heater is optionally available with which the temperature range can be extended to +100°C.
- Minichillers are available as both air and water cooled models and use natural refrigerants.
Spezifikation
Spezifikation
| Kommunikation | USB (Device), RS232 |
| Kühlleistung | 1 kW at 15°C |
| Pumpentyp | Pressure-suction Circulation Pump |
| Temperaturbereich | -20°C to 40°C |
| Typ | Umlaufkühler |
| Modell | Minichiller 1000 OLE |
| Kältemittel | Natürliches Kältemittel |
| Elektrische Anforderungen | 208 to 240 V, 50/60 Hz |
| Spannung | 208/240 V |