Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 555 Antibody Labeling Kit
Antibody labeling kit
Marke: Novus Biologicals™ 333-0010
Weitere Details : Netto-Gewicht : 0.58200kg
Beschreibung
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- 1 vial of LL Rapid Modifier reagent
- 1 vial of LL Rapid Quencher reagent
- Lightning-Link™ Rapid mix
Spezifikation
300 μg | |
Antikörpermarkierungskit | |
-20 °C |