missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF21B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marke: Novus Biologicals NBP1-84188-25ul
Weitere Details : Netto-Gewicht : 0.00970kg
Beschreibung
KIF21B Polyclonal specifically detects KIF21B in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
KIF21B | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
FLJ16314, KIAA0449, kinesin family member 21B | |
Rabbit | |
Affinity Purified | |
RUO | |
23046 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
KIF21B | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RKTQEVSALRRLAKPMSERVAGRAGLKPPMLDSGAEVSASTTSSEAESGARSVSSIVRQWNRKINHFLGDHPAPTVNGTRPARKKFQKKG | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only