Learn More
Oakton ELECTRODE FLUORIDE REFILL BNC
Save money and frustration with the Combination Ion-Selective Electrode (ISE), Fluoride (F) - cost effective and easy to use.
Marke: Oakton 27504-14
304.14 EUR gültig bis 2025-04-30
Benutzen Sie den Promotions-Code "24119" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Easily fit into small spaces—no additional reference electrode is needed.
- These fast and accurate electrodes do not require an external reference—they're easier to use than separate sensing and reference half-cell electrodes.
- Double-junction combination ion-selective electrodes and are convenient to use in places where two electrodes won't fit.
- All electrodes include a 30 mL bottle of reference solution, a reference filling pipette, 1 m cable, BNC connector, and an instruction manual.
- Solid-state electrodes also have twelve polishing strips.
- Economy Epoxy Electrodes have a durable epoxy body and are well-suited for portable field applications.
- Outer reference chamber is silver-free and resists contamination.
- These electrodes are refillable.
- Laboratory Glass Electrodes are double-junction in design, with a glass body that provides high-quality measurements for your laboratory applications.
- Improved glass seal enhances performance and reliability.
- Sealed inner junctions and refillable outer junctions accommodate special reference fill solutions—ideal for low-concentration measurements.
- Use electrodes with a meter that has a resolution of 0.1 mV for the most accurate results.
Spezifikation
BNC-Anschluss | |
Ionen-selektive Elektroden | |
Kombinations-ISE |
Sonderangebote
- Mehrere Hersteller
Berichtigung von Produktinhalten
Name des Produkts
Oakton ELECTRODE FLUORIDE REFILL BNC > Stecker:BNC-Anschluss
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur