missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Hei-CHILL 5000 Base Cart
The Hei-CHILL 5000 Base Cart is the most compact and strongest undercounter chiller with a cooling capacity of 5000 W.
Marke: Heidolph™ 591-01720-01-0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Optimally designed as space-saving solution to be used with a large-scale evaporator and other peripheral devices, this compact undercounter chiller can be perfectly placed among one of our Hei-VAP Industrial models
- The power supply of the rotary evaporator is ensured by a power socket at the rear of the chiller
- Despite its small footprint, this chiller also provides space for a vacuum pump and mounting possibilities for the automatic module Hei-VOLUME Distimatic Pro
- With its cooling capacity of up to 5 kW, this recirculation chiller is extremely powerful
- Its large reservoir is particularly useful for peak loads, and a high flow rate
- Suitable even for demanding liquids like low-boilers and continuous evaporation, combined with the automatic module Hei-VOLUME Distimatic Pro
Spezifikation
28 L | |
Water-cooled | |
40 L/min. | |
Circulation Pump | |
Umlaufkühler | |
208 V |
Keine | |
5000 W (at 20°C), 3500 W (10°C), 2500 W (0°C), 1100 W (-10°C) | |
Large-scale evaporators and other peripheral devices | |
-10°C to 40°C | |
208 V, 50 Hz |