missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-CHILL 5000 Base Cart

The Hei-CHILL 5000 Base Cart is the most compact and strongest undercounter chiller with a cooling capacity of 5000 W.

Marke:  Heidolph™ 591-01720-00-0

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 30057379

  • 16515.00 € / 1 Stück

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

  • Optimally designed as space-saving solution to be used with a large-scale evaporator and other peripheral devices, this compact undercounter chiller can be perfectly placed among one of our Hei-VAP Industrial models
  • The power supply of the rotary evaporator is ensured by a power socket at the rear of the chiller
  • Despite its small footprint, this chiller also provides space for a vacuum pump and mounting possibilities for the automatic module Hei-VOLUME Distimatic Pro
  • With its cooling capacity of up to 5 kW, this recirculation chiller is extremely powerful
  • Its large reservoir is particularly useful for peak loads, and a high flow rate
  • Suitable even for demanding liquids like low-boilers and continuous evaporation, combined with the automatic module Hei-VOLUME Distimatic Pro
Spezifikation

Spezifikation

Keine
5000 W (at 20°C), 3500 W (10°C), 2500 W (0°C), 1100 W (-10°C)
Large-scale evaporators and other peripheral devices
-10°C to 40°C
400 V, 50 Hz
Water-cooled
40 L/min.
Circulation Pump
Umlaufkühler
400 V
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote