Learn More
Eicosapentaenoic Acid, MedChemExpress
MedChemExpress Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA).
Marke: Medchem Express HY-B0660
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Eicosapentaenoic Acid exhibits a DNA demethylating action that promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). Eicosapentaenoic Acid activates RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells. Eicosapentaenoic Acid can promote relaxation of vascular smooth muscle cells and vasodilation.Spezifikation
Eicosapentaenoic Acid | |
Colorless | |
C20H30O2 | |
EPA Timnodonic acid | |
CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O | |
302.45 | |
Research | |
Cancer-programmed cell death |
10417-94-4 | |
Research | |
500 mg | |
Ethanol : ≥ 100 mg/mL (330.63 mM) ∣DMSO : ≥ 53 mg/mL (175.24 mM) | |
302.45 | |
98.0% | |